siRNA Details
Gene Symbol: St8sia5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013666.2 | NP_038694.2 | 3 | 514 | ||
NM_153124.2 | NP_694764.2 | 2 | 406 | |||
GenBank mRNA | AK013542.1 | - | 2 | 409 | ||
AK078670.1 | - | 2 | 189 | |||
AK162549.1 | - | 2 | 186 | |||
BC034855.1 | - | 2 | 400 | 5146703 | ||
BY254855.1 | - | 2 | 410 | |||
BY733136.1 | - | 3 | 496 | |||
X98014.1 | - | 3 | 377 |
