siRNA Details
Gene Symbol: G3bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013716.2 | NP_038744.1 | 4 | 264 | ||
GenBank mRNA | AB001927.1 | - | 4 | 264 | ||
AK137344.1 | - | 4 | 280 | |||
AK141689.1 | - | 4 | 331 | |||
AK179797.1 | - | 3 | 117 | |||
AK220230.1 | - | 4 | 318 | |||
BC021156.1 | - | 4 | 276 | 4020362 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_133565, FQ217639, FQ217678.
Gene Ontology Categories:
