siRNA Details
Gene Symbol: Pvalb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013645.3 | NP_038673.2 | 4 | 535 | ||
GenBank mRNA | AK005371.1 | - | 4 | 533 | ||
AK013561.1 | - | 4 | 527 | |||
X54613.1 | - | 4 | 513 | |||
X59382.1 | - | 5 | 569 | |||
X67141.1 | - | 1 | 210 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
