siRNA Details
Gene Symbol: Rtn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013648.6 | NP_038676.1 | 3 | 395 | ||
GenBank mRNA | AF038538.1 | - | 3 | 351 | ||
AK144271.1 | - | 3 | 358 | |||
BC031370.1 | - | 3 | 352 | 5359358 | ||
CJ176292.1 | - | 3 | 362 |

More Information
Other Species Targeted

Gene Ontology Categories:
