siRNA Details
Gene Symbol: Srsf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013663.5 | NP_038691.1 | 6 | 625 | ||
GenBank mRNA | AK009895.1 | - | 6 | 613 | ||
AK078891.1 | - | 6 | 613 | |||
AK088122.1 | - | 6 | 611 | |||
AK134177.1 | - | 6 | 613 | |||
AK135298.1 | - | 6 | 613 | |||
AK141440.1 | - | 4 | 3156 | |||
AK145089.1 | - | 6 | 614 | |||
AK145600.1 | - | 6 | 613 | |||
AK145715.1 | - | 6 | 613 | |||
AK146010.1 | - | 6 | 613 | |||
AK146253.1 | - | 6 | 611 | |||
AK146610.1 | - | 6 | 613 | |||
AK150912.1 | - | 6 | 613 | |||
AK152411.1 | - | 7 | 1069 | |||
AK152704.1 | - | 6 | 613 | |||
AK152779.1 | - | 7 | 1069 | |||
AK153073.1 | - | 7 | 1069 | |||
AK153209.1 | - | 6 | 613 | |||
AK154046.1 | - | 6 | 613 | |||
BC068111.1 | - | 6 | 605 | 5710934 | ||
BC071196.1 | - | 6 | 587 | 30441615 | ||
BC083316.1 | - | 6 | 597 | 6818630 | ||
X53824.1 | - | 6 | 558 |

More Information
Other Species Targeted

Gene Ontology Categories:
