siRNA Details
Gene Symbol: Tek | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290549.1 | NP_001277478.1 | 23 | 4009 | ||
NM_001290551.1 | NP_001277480.1 | 22 | 3859 | |||
NM_013690.3 | NP_038718.2 | 23 | 4012 | |||
GenBank mRNA | BC050824.1 | - | 22 | 3853 | 6333576 | |
D13738.1 | - | 23 | 4016 | |||
S67051.1 | - | 24 | 3672 | |||
X67553.1 | - | 23 | 3778 |
