siRNA Details
Gene Symbol: Azi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048146.2 | NP_001041611.1 | 2 | 602 | ||
NM_001286507.1 | NP_001273436.1 | 2 | 602 | |||
NM_001286508.1 | NP_001273437.1 | 2 | 520 | |||
NM_013727.4 | NP_038755.1 | 2 | 602 | |||
GenBank mRNA | AB007141.1 | - | 2 | 438 | ||
AK004992.1 | - | 2 | 581 | |||
AK015942.1 | - | 2 | 423 | |||
AK030076.1 | - | 2 | 434 | |||
AK036102.1 | - | 2 | 509 | |||
AK134714.1 | - | 2 | 505 | |||
AK153477.1 | - | 2 | 566 | |||
AK165549.1 | - | 2 | 488 | |||
AK167783.1 | - | 2 | 546 | |||
BC006851.1 | - | 2 | 497 | 3156857 |
