siRNA Details
Gene Symbol: Magel2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013779.2 | NP_038807.4 | 1 | 2877 | ||
GenBank mRNA | AJ243608.1 | - | 1 | 392 | ||
AK082944.1 | - | 1 | 2850 | |||
AK086725.1 | - | 1 | 2867 | |||
BC054763.1 | - | 1 | 169 | |||
BC062913.1 | - | 3 | 2382 | |||
BC071222.1 | - | 1 | 655 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
