siRNA Details
Gene Symbol: Abcc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013790.2 | NP_038818.2 | 7 | 1105 | ||
GenBank mRNA | AB019003.1 | - | 7 | 1056 | ||
AK050565.1 | - | 7 | 1098 | |||
BC090629.1 | - | 7 | 1062 | 6839816 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
