siRNA Details
Gene Symbol: Csnk1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289898.1 | NP_001276827.1 | 11 | 1483 | ||
NM_013767.6 | NP_038795.3 | 11 | 1487 | |||
GenBank mRNA | AK035978.1 | - | 5 | 583 | ||
BC026127.1 | - | 11 | 1435 | 4010696 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
