siRNA Details
Gene Symbol: Abcc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013790.2 | NP_038818.2 | 13 | 2148 | ||
GenBank mRNA | AB019003.1 | - | 13 | 2099 | ||
BC090629.1 | - | 13 | 2105 | 6839816 |

More Information
Other Species Targeted

Gene Ontology Categories:
