siRNA Details
Gene Symbol: Ate1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029895.3 | NP_001025066.1 | 12 | 2450 | ||
NM_001136054.2 | NP_001129526.1 | 12 | 2182 | |||
NM_001271343.1 | NP_001258272.1 | 12 | 2450 | |||
NM_013799.3 | NP_038827.2 | 12 | 2182 | |||
GenBank mRNA | AK077471.1 | - | 8 | 1601 | ||
AK148442.1 | - | 12 | 2150 | |||
AK167591.1 | - | 12 | 2145 | |||
BC042601.1 | - | 12 | 2141 | 4503326 | ||
BC050948.1 | - | 12 | 2132 | 6826691 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
