siRNA Details
Gene Symbol: Zfp957 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033215.3 | NP_001028387.1 | 3 | 2583 | ||
GenBank mRNA | AK139686.1 | - | 3 | 2514 | ||
BC151187.1 | - | 2 | 2177 | 9088076 |
Gene Symbol: Tcf20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114140.1 | NP_001107612.1 | 6 | 6373 | ||
NM_013836.3 | NP_038864.3 | 5 | 6250 | |||
GenBank mRNA | AK122239.1 | - | 5 | 5402 | ||
AK152165.1 | - | 4 | 1429 | |||
BC042485.1 | - | 6 | 3752 | |||
BC080855.1 | - | 4 | 3376 | |||
U20282.1 | - | 4 | 3743 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
