siRNA Details
Gene Symbol: Zfp292 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013889.2 | NP_038917.2 | 8 | 1863 | ||
GenBank mRNA | AB221604.1 | - | 1 | 316 | ||
AF017806.1 | - | 1 | 333 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
