siRNA Details
Gene Symbol: Klra7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110323.1 | NP_001103793.1 | 4 | 497 | ||
NM_014194.5 | NP_055009.5 | 3 | 437 | |||
GenBank mRNA | AK143240.1 | - | 3 | 396 | ||
U10093.1 | - | 4 | 497 | |||
U10094.1 | - | 3 | 437 | |||
U12890.1 | - | 3 | 388 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
