siRNA Details
Gene Symbol: Casp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277932.1 | NP_001264861.1 | 1, 2 | 374 | ||
NM_015733.5 | NP_056548.2 | 1, 2 | 374 | |||
GenBank mRNA | AB019600.1 | - | 1, 2 | 131 | ||
AB019601.1 | - | 1, 2 | 131 | |||
AK085095.1 | - | 1, 2 | 163 | |||
AK085152.1 | - | 1, 2 | 130 | |||
AK085561.1 | - | 1, 2 | 149 | |||
AK154495.1 | - | 1, 2 | 144 | |||
AK156500.1 | - | 1, 2 | 277 | |||
AK162863.1 | - | 1, 2 | 154 | |||
BC056372.1 | - | 1, 2 | 156 | 6834067 | ||
BC056447.1 | - | 1, 2 | 374 | 5721188 | ||
CT010400.1 | - | 1, 2 | 131 |
