siRNA Details
Gene Symbol: Tpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013861.3 | NP_038889.1 | 5, 6 | 321 | ||
GenBank mRNA | AB027568.1 | - | 5, 6 | 322 | ||
AK028431.1 | - | 5, 6 | 344 | |||
AK033192.1 | - | 3, 4 | 425 | |||
AK034521.1 | - | 331 | ||||
AK035044.1 | - | 5, 6 | 322 | |||
AK052225.1 | - | 5, 6 | 313 | |||
AK087844.1 | - | 5, 6 | 425 | |||
AK136486.1 | - | 5, 6 | 322 | |||
BC015246.1 | - | 5, 6 | 338 | 4221866 | ||
BC023354.1 | - | 5, 6 | 297 | 5253545 |
