siRNA Details
Gene Symbol: Heyl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB093590.1 | - | 5 | 1025 | ||
AF172288.1 | - | 5 | 1333 | |||
AJ271868.2 | - | 5 | 1333 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
