siRNA Details
Gene Symbol: Tpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013861.3 | NP_038889.1 | 9 | 2033 | ||
GenBank mRNA | AB027568.1 | - | 9 | 2034 | ||
AK028431.1 | - | 9 | 2055 | |||
AK033192.1 | - | 7 | 2137 | |||
AK034521.1 | - | 2042 | ||||
AK035044.1 | - | 9 | 2034 | |||
AK052225.1 | - | 9 | 2025 | |||
AK136486.1 | - | 8 | 1887 | |||
BC015246.1 | - | 9 | 2050 | 4221866 | ||
BC023354.1 | - | 8 | 1862 | 5253545 |
