siRNA Details
Gene Symbol: Scmh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159630.1 | NP_001153102.1 | 14, 15 | 2126 | ||
NM_013883.2 | NP_038911.1 | 14, 15 | 2126 | |||
GenBank mRNA | AB030906.1 | - | 14, 15 | 2025 | ||
AB221680.1 | - | 10, 11 | 1667 | |||
AB221681.1 | - | 10, 11 | 1667 | |||
AK009416.1 | - | 2, 3 | 284 | |||
BC034667.1 | - | 12, 13 | 1836 | 5356322 |
