siRNA Details
Gene Symbol: Snai3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013914.2 | NP_038942.1 | 2, 3 | 768 | ||
GenBank mRNA | AF133714.1 | - | 2, 3 | 690 | ||
AF195655.1 | - | 2, 3 | 690 | |||
AK042234.1 | - | 2, 3 | 768 | |||
AK089285.1 | - | 2, 3 | 757 | |||
AK153633.1 | - | 2, 3 | 810 | |||
AK154585.1 | - | 1, 2 | 446 | |||
BC100724.1 | - | 2, 3 | 717 | 40045286 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
