siRNA Details
Gene Symbol: Cacna1s | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081023.1 | NP_001074492.1 | 38 | 4631 | ||
NM_014193.2 | NP_055008.2 | 37 | 4574 | |||
GenBank mRNA | AK142239.1 | - | 32 | 3833 | ||
AK143541.1 | - | 37 | 4512 | |||
BC134369.1 | - | 6 | 455 | |||
L06234.1 | - | 37 | 4689 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
