siRNA Details
Gene Symbol: Casp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277932.1 | NP_001264861.1 | 8 | 1438 | ||
NM_015733.5 | NP_056548.2 | 9 | 1548 | |||
GenBank mRNA | AB019600.1 | - | 9 | 1305 | ||
AB019601.1 | - | 8 | 1195 | |||
AK085095.1 | - | 9 | 1337 | |||
AK085152.1 | - | 9 | 1304 | |||
AK154495.1 | - | 9 | 1318 | |||
BC056372.1 | - | 9 | 1330 | 6834067 | ||
BC056447.1 | - | 9 | 1548 | 5721188 | ||
CT010400.1 | - | 9 | 1305 |
