siRNA Details
Gene Symbol: Hspa14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015765.2 | NP_056580.2 | 5 | 445 | ||
GenBank mRNA | AK031489.1 | - | 384 | |||
AK145937.1 | - | 5 | 399 | |||
AK153397.1 | - | 5 | 407 | |||
BC002056.1 | - | 5 | 373 | 3493119 | ||
BU505607.1 | - | 2 | 96 | |||
U08215.1 | - | 5 | 405 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
