siRNA Details
Gene Symbol: TNK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010938.1 | NP_001010938.1 | 3 | 539 | ||
NM_001308046.1 | NP_001294975.1 | 3 | 597 | |||
NM_005781.4 | NP_005772.3 | 3 | 748 | |||
XM_005269268.3 | XP_005269325.1 | 3 | 539 | |||
XM_005269270.3 | XP_005269327.1 | 3 | 387 | |||
XM_005269275.3 | XP_005269332.1 | 3 | 561 | |||
XM_011512317.2 | XP_011510619.1 | 5 | 1106 | |||
XM_011512318.2 | XP_011510620.2 | 3 | 478 | |||
XM_011512320.1 | XP_011510622.1 | 3 | 261 | |||
XM_017005508.1 | XP_016860997.1 | 3 | 478 | |||
XM_017005509.1 | XP_016860998.1 | 3 | 478 | |||
XM_017005510.1 | XP_016860999.1 | 3 | 478 | |||
GenBank mRNA | AK131539.1 | - | 3 | 539 | ||
AK225786.1 | - | 3 | 544 | |||
AK309332.1 | - | 3 | 375 | |||
BC008884.2 | - | 3 | 394 | |||
BC020389.1 | - | 3 | 588 | |||
BC028164.1 | - | 3 | 262 | |||
L13738.2 | - | 3 | 823 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
