siRNA Details
Gene Symbol: Dlc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001194940.2 | NP_001181869.1 | 9 | 2069 | ||
NM_001194941.1 | NP_001181870.1 | 5 | 699 | |||
NM_015802.3 | NP_056617.2 | 5 | 617 | |||
GenBank mRNA | AF178078.1 | - | 5 | 553 | ||
AK147539.1 | - | 5 | 699 | |||
AK162413.1 | - | 5 | 618 | |||
AK166175.1 | - | 5 | 577 | |||
BC150802.1 | - | 5 | 402 | 9087713 | ||
GU169069.1 | - | 6 | 323 | |||
HM008381.1 | - | 8 | 1622 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
