siRNA Details
Gene Symbol: Atp9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201569.1 | NP_001188498.1 | 2 | 371 | ||
NM_015805.3 | NP_056620.2 | 2 | 371 | |||
GenBank mRNA | AB206122.1 | - | 1 | 99 | ||
AF155913.1 | - | 1 | 99 | |||
AK030485.1 | - | 1 | 32 | |||
AK044331.1 | - | 2 | 346 | |||
AK166562.1 | - | 2 | 374 | |||
BC057000.1 | - | 2 | 296 | |||
BC079626.1 | - | 2 | 296 | 30536500 | ||
BY735600.1 | - | 2 | 374 |

More Information
Other Species Targeted

Gene Ontology Categories:
