siRNA Details
Gene Symbol: Hspa14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015765.2 | NP_056580.2 | 8, 9 | 866 | ||
GenBank mRNA | AK031489.1 | - | 805 | |||
AK145937.1 | - | 8, 9 | 820 | |||
AK153397.1 | - | 8, 9 | 828 | |||
BC002056.1 | - | 8, 9 | 794 | 3493119 | ||
BU505607.1 | - | 5, 6 | 517 | |||
U08215.1 | - | 8, 9 | 826 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
