siRNA Details
Gene Symbol: Prok2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015768.2 | NP_056583.1 | 3 | 319 | ||
GenBank mRNA | AF182064.1 | - | 3 | 319 | ||
AK015462.1 | - | 3 | 281 | |||
BC116911.1 | - | 3 | 288 | 40126230 |

More Information
Other Species Targeted

Gene Ontology Categories:
