siRNA Details
Gene Symbol: Atp9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201569.1 | NP_001188498.1 | 3 | 450 | ||
NM_015805.3 | NP_056620.2 | 3 | 450 | |||
GenBank mRNA | AB206122.1 | - | 2 | 178 | ||
AF155913.1 | - | 2 | 178 | |||
AK030485.1 | - | 2 | 111 | |||
AK044331.1 | - | 3 | 425 | |||
AK166562.1 | - | 3 | 453 | |||
BC057000.1 | - | 3 | 375 | |||
BC079626.1 | - | 3 | 375 | 30536500 | ||
BY735600.1 | - | 3 | 453 |
