siRNA Details
Gene Symbol: Dlc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001194940.2 | NP_001181869.1 | 13, 14 | 4240 | ||
NM_001194941.1 | NP_001181870.1 | 9, 10 | 2870 | |||
NM_015802.3 | NP_056617.2 | 9, 10 | 2788 | |||
GenBank mRNA | AF178078.1 | - | 9, 10 | 2724 | ||
AK147539.1 | - | 9, 10 | 2870 | |||
AK162413.1 | - | 9, 10 | 2789 | |||
AK166175.1 | - | 9, 10 | 2748 | |||
BC150802.1 | - | 9, 10 | 2573 | 9087713 | ||
HM008381.1 | - | 12, 13 | 3793 |

More Information
Other Species Targeted

Gene Ontology Categories:
