siRNA Details
Gene Symbol: Atp9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201569.1 | NP_001188498.1 | 5, 6 | 791 | ||
NM_015805.3 | NP_056620.2 | 5, 6 | 791 | |||
GenBank mRNA | AB206122.1 | - | 4, 5 | 519 | ||
AF155913.1 | - | 4, 5 | 519 | |||
AK030485.1 | - | 4, 5 | 452 | |||
AK044331.1 | - | 5, 6 | 766 | |||
BC057000.1 | - | 5, 6 | 716 | |||
BC079626.1 | - | 5, 6 | 716 | 30536500 |

More Information
Other Species Targeted

Gene Ontology Categories:
