siRNA Details
Gene Symbol: Hs6st1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015818.2 | NP_056633.2 | 2 | 3611 | ||
GenBank mRNA | AB024566.1 | - | 2 | 3602 | ||
AK009411.1 | - | 1 | 134 | |||
AK166962.1 | - | 2716 | ||||
AK167151.1 | - | 2 | 3040 | |||
BC052316.1 | - | 2 | 3611 | 6391204 |
