siRNA Details
Gene Symbol: Rundc3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252347.1 | NP_001239276.1 | 1, 2 | 388 | ||
NM_016759.4 | NP_058039.1 | 1, 2 | 388 | |||
GenBank mRNA | BC046319.1 | - | 1, 2 | 288 | 6391042 | |
BY258909.1 | - | 1, 2 | 361 | |||
BY277835.1 | - | 1, 2 | 389 | |||
U73941.1 | - | 1, 2 | 115 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
