siRNA Details
Gene Symbol: Srpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016795.3 | NP_058075.2 | 2, 3 | 121 | ||
GenBank mRNA | AB012290.1 | - | 1, 2 | 79 | ||
AJ224115.1 | - | 2, 3 | 115 | |||
AK017187.1 | - | 2, 3 | 118 | |||
AK141271.1 | - | 2, 3 | 188 | |||
AK151152.1 | - | 2, 3 | 184 | |||
AK161346.1 | - | 2, 3 | 99 | |||
AK161388.1 | - | 2, 3 | 100 | |||
BC005707.1 | - | 2, 3 | 90 | 3157835 | ||
BC050761.1 | - | 2, 3 | 121 | 6771151 |
