siRNA Details
Gene Symbol: Incenp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016692.3 | NP_057901.2 | 2 | 319 | ||
GenBank mRNA | AB100432.1 | - | 1 | 113 | ||
AB100433.1 | - | 1 | 113 | |||
AF117610.1 | - | 2 | 255 | |||
AK034764.1 | - | 2 | 248 | |||
AK081841.1 | - | 2 | 253 | |||
AK088627.1 | - | 2 | 276 | |||
BC021761.2 | - | 2 | 216 | |||
BC037011.1 | - | 2 | 272 | |||
BC052414.1 | - | 2 | 252 | 6401188 |
