siRNA Details
Gene Symbol: Pgrmc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016783.3 | NP_058063.2 | 2 | 478 | ||
GenBank mRNA | AF042491.1 | - | 2 | 429 | ||
AK153272.1 | - | 2 | 495 | |||
AK159101.1 | - | 2 | 495 | |||
AK159345.1 | - | 2 | 495 | |||
AK159599.1 | - | 2 | 495 | |||
AK162315.1 | - | 2 | 498 | |||
AK167655.1 | - | 2 | 495 | |||
AK168387.1 | - | 2 | 494 | |||
AK168882.1 | - | 2 | 495 | |||
AK169063.1 | - | 3 | 438 | |||
AK193145.1 | - | 1 | 66 | |||
AK208626.1 | - | 1 | 66 | |||
AK214321.1 | - | 1 | 66 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
