siRNA Details
Gene Symbol: Nfatc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164109.1 | NP_001157581.1 | 8 | 2453 | ||
NM_016791.4 | NP_058071.2 | 8 | 2161 | |||
GenBank mRNA | AF049606.1 | - | 8 | 2114 | ||
AF087434.1 | - | 8 | 2148 | |||
AF239169.1 | - | 8 | 2151 | |||
AK004810.1 | - | 8 | 2119 | |||
AK048610.1 | - | 8 | 2455 | |||
EU887570.1 | - | 8 | 2148 | |||
EU887571.1 | - | 8 | 2106 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
