siRNA Details
Gene Symbol: Nptx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016789.3 | NP_058069.1 | 5 | 1521 | ||
GenBank mRNA | AF049124.1 | - | 5 | 1411 | ||
AK134806.1 | - | 5 | 1522 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
