siRNA Details
Gene Symbol: Srpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016795.3 | NP_058075.2 | 11 | 1184 | ||
GenBank mRNA | AB012290.1 | - | 10 | 1142 | ||
AJ224115.1 | - | 11 | 1178 | |||
AK015754.1 | - | 7 | 712 | |||
AK140761.1 | - | 594 | ||||
AK151152.1 | - | 11 | 1247 | |||
AK164733.1 | - | 1 | 1169 | |||
BC005707.1 | - | 11 | 1153 | 3157835 | ||
BC050761.1 | - | 11 | 1184 | 6771151 |
