siRNA Details
Gene Symbol: Fkbp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016863.3 | NP_058559.3 | 2, 3 | 253 | ||
GenBank mRNA | AB107918.1 | - | 2, 3 | 93 | ||
AB285017.1 | - | 2, 3 | 181 | |||
AF060872.1 | - | 2, 3 | 197 | |||
AK005201.1 | - | 2, 3 | 255 | |||
BC061121.1 | - | 2, 3 | 231 | 6535176 | ||
BE861051.1 | - | 2, 3 | 268 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
