siRNA Details
Gene Symbol: PPIH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006347.3 | NP_006338.1 | 4, 5 | 278 | ||
XM_005270362.1 | XP_005270419.1 | 4, 5 | 232 | |||
XM_005270366.3 | XP_005270423.1 | 4, 5 | 885 | |||
XM_011540508.1 | XP_011538810.1 | 4, 5 | 350 | |||
XM_017000057.1 | XP_016855546.1 | 4, 5 | 232 | |||
XM_017000058.1 | XP_016855547.1 | 4, 5 | 1004 | |||
XM_017000059.1 | XP_016855548.1 | 4, 5 | 350 | |||
GenBank mRNA | AF016371.1 | - | 4, 5 | 245 | ||
AF036331.1 | - | 4, 5 | 221 | |||
AK294288.1 | - | 2, 3 | 507 | |||
AK309438.1 | - | 4, 5 | 346 | |||
AK310052.1 | - | 3, 4 | 501 | |||
AK311979.1 | - | 4, 5 | 231 | |||
BC003412.1 | - | 4, 5 | 230 | 3451034 | ||
BT019343.1 | - | 4, 5 | 208 | |||
BU955702.1 | - | 4, 5 | 287 | |||
CR541899.1 | - | 4, 5 | 208 | |||
CR541920.1 | - | 4, 5 | 208 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
