siRNA Details
Gene Symbol: BCKDK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122957.2 | NP_001116429.1 | 4 | 673 | ||
NM_001271926.1 | NP_001258855.1 | 4 | 673 | |||
NM_005881.3 | NP_005872.2 | 4 | 673 | |||
XM_017022859.1 | XP_016878348.1 | 4 | 586 | |||
GenBank mRNA | AF026548.1 | - | 4 | 583 | ||
AK130145.1 | - | 4 | 553 | |||
AK294691.1 | - | 3 | 382 | |||
BC007363.2 | - | 4 | 556 | 3630050 | ||
BC009872.2 | - | 4 | 536 | 3944953 | ||
BX427330.2 | - | 4 | 670 | |||
CR542093.1 | - | 3 | 310 |

More Information
Other Species Targeted

Gene Ontology Categories:
