siRNA Details
Gene Symbol: Hnrnpu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016805.2 | NP_058085.2 | 2 | 864 | ||
GenBank mRNA | AF073992.1 | - | 2 | 801 | ||
AK029944.1 | - | 2 | 810 | |||
AK037517.1 | - | 2 | 806 | |||
AK089058.1 | - | 2 | 865 | |||
AK145499.1 | - | 2 | 867 | |||
AK145682.1 | - | 2 | 866 | |||
AK159084.1 | - | 2 | 833 | |||
AK159614.1 | - | 2 | 869 | |||
AK159956.1 | - | 2 | 806 | |||
AK162264.1 | - | 2 | 869 | |||
AK165844.1 | - | 2 | 864 | |||
AK167200.1 | - | 2 | 866 | |||
AK168657.1 | - | 2 | 806 | |||
BC018353.1 | - | 2 | 709 | 3584692 |
