siRNA Details
Gene Symbol: Spint1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016907.3 | NP_058603.2 | 11 | 2151 | ||
GenBank mRNA | AF099018.1 | - | 11 | 2106 | ||
AK017342.1 | - | 10 | 2033 | |||
BC005769.1 | - | 10 | 2038 | 2650110 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
