siRNA Details
Gene Symbol: Hmgn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK146285.1 | - | 3, 4 | 241 | ||
AK153881.1 | - | 186 | ||||
BC084680.1 | - | 3, 4 | 122 | 6477991 | ||
BC103792.1 | - | 170 | 6516212 | |||
X12944.1 | - | 127 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001025624, XM_003751587, FQ216489, BC099815, FQ213300, and others.
