siRNA Details
Gene Symbol: Rfx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017395.2 | NP_059091.2 | 1 | 122 | ||
GenBank mRNA | AB221679.1 | - | 1 | 103 | ||
AF209854.1 | - | 1 | 103 | |||
AK088686.1 | - | 2 | 174 | |||
BB854843.1 | - | 2 | 215 | |||
BC057336.1 | - | 2 | 148 | |||
BC075717.1 | - | 3 | 231 | |||
BC118948.2 | - | 1 | 103 | 40092396 | ||
BC118949.2 | - | 1 | 103 | 40092397 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
