siRNA Details
Gene Symbol: IGF1R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000875.4 | NP_000866.1 | 2 | 1429 | ||
NM_001291858.1 | NP_001278787.1 | 2 | 1429 | |||
XM_017022136.1 | XP_016877625.1 | 2 | 932 | |||
XM_017022137.1 | XP_016877626.1 | 2 | 932 | |||
XM_017022138.1 | XP_016877627.1 | 2 | 932 | |||
XM_017022139.1 | XP_016877628.1 | 2 | 370 | |||
GenBank mRNA | AB425196.1 | - | 2 | 915 | ||
BC113610.1 | - | 2 | 418 | 8322662 | ||
BC113612.1 | - | 2 | 418 | 8322664 | ||
BC143721.1 | - | 2 | 418 | 9052235 | ||
EU826611.1 | - | 2 | 386 | |||
EU826612.1 | - | 2 | 386 | |||
X04434.1 | - | 2 | 431 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
