siRNA Details
Gene Symbol: Icos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017480.2 | NP_059508.2 | 2 | 162 | ||
GenBank mRNA | AB023132.1 | - | 2 | 146 | ||
AF216748.1 | - | 2 | 117 | |||
AF257230.1 | - | 2 | 147 | |||
AJ250559.1 | - | 2 | 162 | |||
AK030827.1 | - | 2 | 155 | |||
BB850356.1 | - | 2 | 163 | |||
BC034852.1 | - | 2 | 149 | 1364228 |

More Information
Other Species Targeted

Gene Ontology Categories:
