siRNA Details
Gene Symbol: Gbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289492.1 | NP_001276421.1 | 3 | 532 | ||
NM_001289493.1 | NP_001276422.1 | 2 | 357 | |||
NM_018734.3 | NP_061204.3 | 2 | 394 | |||
GenBank mRNA | AK132266.1 | - | 2 | 393 | ||
CK389037.1 | - | 2 | 371 | |||
U44731.1 | - | 2 | 246 |

More Information
Other Species Targeted

Gene Ontology Categories:
